ENAH5802EEB8 Enamine

N-(propan-2-yl)thiophene-2-carbonimidoyl chloride

  • Cas Number : 885524-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14849
Availability details for
Quantity
  • Availability
  • Add to Cart