ENAH5802EED1 Enamine

(3aR,6aR)-2-imino-hexahydro-5lambda6-thieno[3,4-d][1,3]thiazole-5,5-dione

  • MDL number MFCD31566993
  • Cas Number : 1808655-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207414
Availability details for
Quantity
  • Availability
  • Add to Cart