ENAH5802EEE2 Enamine

lithium(1+) ion 3-(but-3-en-1-yl)benzene-1-sulfinate

  • MDL number MFCD32197701
  • Cas Number : 2375267-91-5
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7438029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438029
Availability details for
Quantity
  • Availability
  • Add to Cart