ENAH5802EEF0 Enamine

3-(5-methylthiophen-2-yl)-2,5-dihydro-1,2,4-thiadiazol-5-imine

  • MDL number MFCD26143849
  • Cas Number : 942890-87-1
  • Molecular Weight : 197.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65587
Availability details for
Quantity
  • Availability
  • Add to Cart