ENAH5802EF10 Enamine

1H,3H,4H,6H-morpholino[3,4-b]quinazolin-6-one

  • MDL number MFCD31924309
  • Cas Number : 19650-95-4
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7353860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353860
Availability details for
Quantity
  • Availability
  • Add to Cart