ENAH5802EF1E Enamine

4-[3-(propan-2-yl)-2,5-dihydro-1,2,4-oxadiazol-5-ylidene]cyclohexa-2,5-dien-1-imine

  • MDL number MFCD26143797
  • Cas Number : 1955541-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64580
Availability details for
Quantity
  • Availability
  • Add to Cart