ENAH5802EF2D Enamine

4-chloro-1-methyl-3,5-dinitro-1H-pyrazole

  • MDL number MFCD00463991
  • Cas Number : 84547-96-6
  • Molecular Weight : 206.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426228
Availability details for
Quantity
  • Availability
  • Add to Cart