ENAH5802EF36 Enamine

4-(3-chlorophenyl)-3-methyl-2,5-dihydro-1,2-oxazol-5-imine

  • MDL number MFCD26143817
  • Cas Number : 1311856-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73367
Availability details for
Quantity
  • Availability
  • Add to Cart