ENAH5802EF46 Enamine

4-(1H-1,3-benzodiazol-2-yl)-2-methyl-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD26143694
  • Cas Number : 1955524-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26387
Availability details for
Quantity
  • Availability
  • Add to Cart