ENAH5802EF47 Enamine

N-(2-methylpropyl)benzenesulfonamide

  • MDL number MFCD00956408
  • Cas Number : 23705-39-7
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7435543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435543
Availability details for
Quantity
  • Availability
  • Add to Cart