ENAH5802EF51 Enamine

2-tert-butyl-5-phenyl-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD28155307
  • Cas Number : 1955558-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76503
Availability details for
Quantity
  • Availability
  • Add to Cart