ENAH5802EF55 Enamine

1,1,2,3,3-pentachloropropane

  • MDL number MFCD00018862
  • Cas Number : 15104-61-7
  • Molecular Weight : 216.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-392956

Safety Information

signalword Warning
pcodes P210,P261,P264,P270,P271,P273,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P370+P378,P391,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-392956
pictograms GHS07,GHS09
hstatements H227,H302,H312,H315,H319,H332,H411
Availability details for
Quantity
  • Availability
  • Add to Cart