ENAH5802EF74 Enamine

3-[(4-methylphenyl)sulfanyl]-2,5-dihydro-1,2,4-thiadiazol-5-imine

  • MDL number MFCD28155329
  • Cas Number : 1256627-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92936
Availability details for
Quantity
  • Availability
  • Add to Cart