ENAH5802EF76 Enamine

ethyl 4-(1-methyl-1H-pyrazol-4-yl)-2,4-dioxobutanoate

  • MDL number MFCD16109055
  • Cas Number : 873221-73-9
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1609256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1609256
Availability details for
Quantity
  • Availability
  • Add to Cart