ENAH5802EF7F Enamine

1-(2-methylphenyl)-1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD26143775
  • Cas Number : 1955505-85-7
  • Molecular Weight : 226.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(o-tolyl)-1,2-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
Inchi Code 1S/C12H10N4O/c1-8-4-2-3-5-10(8)16-11-9(6-15-16)12(17)14-7-13-11/h2-7,15H,1H3
Inchi Key HAXUMFFHYHMZFR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-78518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78518
Availability details for
Quantity
  • Availability
  • Add to Cart