ENAH5802EF87 Enamine

2-(chloromethyl)-5,6-dimethyl-1H,4H-thieno[2,3-d]pyrimidin-4-imine

  • MDL number MFCD26143668
  • Cas Number : 877964-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23963
Availability details for
Quantity
  • Availability
  • Add to Cart