ENAH5802EFA6 Enamine

5-methyl-N-[4-(propan-2-yl)phenyl]-1,3-thiazolidin-2-imine

  • Cas Number : 721418-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-04312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-04312
Availability details for
Quantity
  • Availability
  • Add to Cart