ENAH5802EFB3 Enamine

5-(furan-2-yl)-2-(4-methylphenyl)-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD23146396
  • Cas Number : 1333887-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81969
Availability details for
Quantity
  • Availability
  • Add to Cart