ENAH5802EFB5 Enamine

ethyl 2-(2-ethoxy-2-oxoethyl)-3-imino-2,3-dihydro-1H-pyrazole-4-carboxylate

  • MDL number MFCD28155346
  • Cas Number : 1356754-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70822
Availability details for
Quantity
  • Availability
  • Add to Cart