ENAH5802EFBF Enamine

1-phenyl-3-[2-(propan-2-yl)-2,3-dihydro-1H-pyrazol-3-ylidene]urea

  • Cas Number : 1955564-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24248
Availability details for
Quantity
  • Availability
  • Add to Cart