ENAH5802EFC1 Enamine

4-(1,3-dioxaindan-5-yl)-5-ethyl-2-methyl-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD28155335
  • Cas Number : 1355666-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-93924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93924
Availability details for
Quantity
  • Availability
  • Add to Cart