ENAH5802EFC4 Enamine

1-(4-fluorophenyl)-1H,2H,4H-pyrazolo[3,4-d]pyrimidine-4-thione

  • MDL number MFCD26143681
  • Cas Number : 1955554-42-3
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25646
Availability details for
Quantity
  • Availability
  • Add to Cart