ENAH5802EFDA Enamine

2-(3,5-dimethylphenyl)-5-(furan-2-yl)-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD23146397
  • Cas Number : 1334015-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-80877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80877
Availability details for
Quantity
  • Availability
  • Add to Cart