ENAH5802EFDD Enamine

2-(3,4-dichlorophenyl)-4,5-dimethyl-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD28155320
  • Cas Number : 1356744-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-80333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80333
Availability details for
Quantity
  • Availability
  • Add to Cart