ENAH5802EFE5 Enamine

1,3-dimethyl-6-(trifluoromethyl)-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-2,4-dione

  • MDL number MFCD32066661
  • Cas Number : 2361634-27-5
  • Molecular Weight : 259.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6763860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763860
Availability details for
Quantity
  • Availability
  • Add to Cart