ENAH5802EFED Enamine

methyl 2-{12-imino-7-thia-9,11-diazatricyclo[6.4.0.0,2,6]dodeca-1(8),2(6),10-trien-10-yl}acetate

  • MDL number MFCD26143684
  • Cas Number : 923769-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25793
Availability details for
Quantity
  • Availability
  • Add to Cart