ENAH5802EFF0 Enamine

2,2,2-trifluoroethyl N-[2-(2-methylpropyl)-2,3-dihydro-1H-pyrazol-3-ylidene]carbamate

  • Cas Number : 1334027-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56687
Availability details for
Quantity
  • Availability
  • Add to Cart