ENAH5802EFFE Enamine

2,3-difluoro-2,3-dimethylbutyl trifluoromethanesulfonate

  • MDL number MFCD31924720
  • Cas Number : 2344681-60-1
  • Molecular Weight : 270.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7424788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7424788
Availability details for
Quantity
  • Availability
  • Add to Cart