ENAH5802F00B Enamine

4-(4-chlorophenyl)-2-methyl-5-(trifluoromethyl)-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD26143819
  • Cas Number : 1365630-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66725
Availability details for
Quantity
  • Availability
  • Add to Cart