ENAH5802F01A Enamine

3-methyl-8-nitro-7-pentyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione

  • MDL number MFCD00616604
  • Cas Number : 203451-40-5
  • Molecular Weight : 281.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7439053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439053
Availability details for
Quantity
  • Availability
  • Add to Cart