ENAH5802F01C Enamine

6-(1-cyclopropyl-6-fluoro-2,3-dihydro-1H-1,3-benzodiazol-2-ylidene)-2-methylcyclohexa-2,4-dien-1-imine

  • MDL number MFCD28657114
  • Cas Number : 1355939-28-4
  • Molecular Weight : 281.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-85934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85934
Availability details for
Quantity
  • Availability
  • Add to Cart