ENAH5802F03A Enamine

1-(2-methylpropyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione

  • MDL number MFCD07841691
  • Cas Number : 7624-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23635
Availability details for
Quantity
  • Availability
  • Add to Cart