ENAH5802F03F Enamine

ethyl N-[(1S,2R)-2-(aminomethyl)-1-(4-methoxyphenyl)-3-methylbutyl]carbamate

  • MDL number MFCD26143870
  • Cas Number : 1808568-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43818
Availability details for
Quantity
  • Availability
  • Add to Cart