ENAH5802F05B Enamine

4-{[5-(3H-indol-3-ylidene)-4,5-dihydro-1,3,4-oxadiazol-2-yl]amino}benzoic acid

  • Cas Number : 1955564-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26911
Availability details for
Quantity
  • Availability
  • Add to Cart