ENAH5802F07A Enamine

tert-butyl 7-[(4-methoxyphenyl)methyl]-3,7-diazatricyclo[3.3.2.0,1,5]decane-3-carboxylate

  • MDL number MFCD31924609
  • Cas Number : 2344685-92-1
  • Molecular Weight : 358.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6740164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740164
Availability details for
Quantity
  • Availability
  • Add to Cart