ENAH5802F0A0 Enamine

N-methylthiophene-2-carbonimidoyl chloride

  • Cas Number : 71267-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14634
Availability details for
Quantity
  • Availability
  • Add to Cart