ENAH5802F0BF Enamine

2-[(1s,3s)-3-aminocyclobutyl]acetonitrile hydrochloride

  • Cas Number : 2361645-64-7
  • Molecular Weight : 146.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432967
Availability details for
Quantity
  • Availability
  • Add to Cart