ENAH5802F0CF Enamine

1-(2-imino-1,2-dihydropyrimidin-4-yl)methanamine

  • MDL number MFCD31583480
  • Cas Number : 929973-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34939
Availability details for
Quantity
  • Availability
  • Add to Cart