ENAH5802F0E6 Enamine

4-(methylsulfanyl)-1H-pyrazol-5-amine

  • MDL number MFCD23718353
  • Cas Number : 1249616-13-4
  • Molecular Weight : 129.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(methylthio)-1H-pyrazol-5-amine
Inchi Code 1S/C4H7N3S/c1-8-3-2-6-7-4(3)5/h2H,1H3,(H3,5,6,7)
Inchi Key MHSNHSVOPUBWGL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-97966

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-97966
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart