ENAH5802F0F0 Enamine

(1s,3s)-3-[(methylsulfanyl)methyl]cyclobutan-1-amine hydrochloride

  • Cas Number : 2361636-01-1
  • Molecular Weight : 167.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7429945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7429945
Availability details for
Quantity
  • Availability
  • Add to Cart