ENAH5802F105 Enamine

2-(1-ethyl-1H-pyrrol-2-yl)ethan-1-amine

  • MDL number MFCD21767988
  • Cas Number : 1354777-29-9
  • Molecular Weight : 138.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2983283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2983283
Availability details for
Quantity
  • Availability
  • Add to Cart