ENAH5802F109 Enamine

4-(2-fluoroethyl)aniline

  • MDL number MFCD19214685
  • Cas Number : 327985-14-8
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4264867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4264867
Availability details for
Quantity
  • Availability
  • Add to Cart