ENAH5802F10A Enamine

5-(1,1-difluoroethyl)-2-fluoropyridine

  • MDL number MFCD32066684
  • Cas Number : 2361636-04-4
  • Molecular Weight : 161.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430147
Availability details for
Quantity
  • Availability
  • Add to Cart