ENAH5802F1A8 Enamine

2-amino-3-(1-methyl-1H-1,2,4-triazol-5-yl)propanamide dihydrochloride

  • MDL number MFCD32198048
  • Cas Number : 2375268-04-3
  • Molecular Weight : 242.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443446
Availability details for
Quantity
  • Availability
  • Add to Cart