ENAH5802F249 Enamine

2',3'-dihydro-1'H-spiro[oxane-4,4'-pyrrolo[1,2-a]pyrazine]

  • MDL number MFCD32069312
  • Cas Number : 2375274-73-8
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430967
Availability details for
Quantity
  • Availability
  • Add to Cart