ENAH5802F299 Enamine

7-(trifluoromethyl)quinolin-2-amine

  • MDL number MFCD11108687
  • Cas Number : 113508-12-6
  • Molecular Weight : 212.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7407709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7407709
Availability details for
Quantity
  • Availability
  • Add to Cart