ENAH5802F2C4 Enamine

4-amino-3-[2-(dimethylamino)ethyl]-2,3-dihydroquinazolin-2-one

  • MDL number MFCD26143682
  • Cas Number : 1955519-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-26181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26181
Availability details for
Quantity
  • Availability
  • Add to Cart