ENAH5802F30E Enamine

5-amino-N-[(4-methoxyphenyl)methyl]-1H-pyrazole-4-sulfonamide

  • MDL number MFCD31583492
  • Cas Number : 1184583-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81601
Availability details for
Quantity
  • Availability
  • Add to Cart