ENAH5802F32D Enamine

3-cyclopropyl-3-methylazetidine

  • MDL number MFCD32066324
  • Cas Number : 1889188-22-0
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1604724

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1604724
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart