ENAH5802F338 Enamine

1H,4H,5H,6H,7H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD11865066
  • Cas Number : 871792-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89809
Availability details for
Quantity
  • Availability
  • Add to Cart